Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

[D-Phe12, Leu14] Bombesin

Supplier: Thermo Scientific Chemicals

J66013-MCR J66013-LB0
10815-372EA 339.49 USD InStock
10815-372 10815-374
[D-Phe12, Leu14] Bombesin
[D-Phe12, Leu14] Bombesin


Formula: C₇₅H₁₁₄N₂₂O₁₈
MW: 1611.873 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00076256
CAS Number: 108437-88-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty