Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Ribose-5-phosphate barium salt hexahydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

J66017-MD J66017-03
10815-378EA 125.8 USD InStock
10815-378 10815-376
D-Ribose-5-phosphate barium salt hexahydrate 99%
D-Ribose-5-phosphate barium salt hexahydrate


Formula: C₅H₁₁O₈P
Storage Temperature: Refrigerator
MDL Number: MFCD00047534
CAS Number: 15673-79-7
EINECS: 239-750-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty