Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Glucose-6-phosphate disodium salt 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Robison Ester Disodium Salt

J66023-03 J66023-06
10815-384EA 70.85 USD InStock
10815-384 10815-386
D-Glucose-6-phosphate disodium salt 98%
D-Glucose-6-phosphate disodium salt


Formula: C₆H₁₁Na₂O₉P
MW: 304.098 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00136037
CAS Number: 3671-99-6
EINECS: 222-938-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty