Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Glucuronamide 98%

Supplier: Thermo Scientific Chemicals

J66047-03 J66047-06
10815-416EA 83.5 USD InStock
10815-416 10815-414
D-Glucuronamide 98%
D-Glucuronamide


Formula: C₆H₁₁NO₆
Melting Pt: 166…167 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006619
CAS Number: 3789-97-7
EINECS: 223-260-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty