Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3,4,6-Tetra-O-acetyl-D-glucopyranose 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 2,3,4,6-tetra-O-acetyl-beta-D-glucopyranose2,3,4,6-tetra-O-acetylglucopyranoseD-Glucopyranose 2,3,4,6-tetraacetate

J66052-06 J66052-03 J66052-09
10815-426EA 201.89 USD InStock
10815-426 10815-428 10815-424
2,3,4,6-Tetra-O-acetyl-D-glucopyranose 98%
2,3,4,6-Tetra-O-acetyl-D-glucopyranose


Formula: C₁₄H₂₀O₁₀
MW: 348.304 g/mol
Melting Pt: 137…138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00061634
CAS Number: 10343-06-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty