Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human [Tyr0] Gastric Inhibitory Peptide

Supplier: Thermo Scientific
J66109-MCR J66109-EXD
10815-444EA 203.92 USD InStock
10815-444 10815-446
Human [Tyr0] Gastric Inhibitory Peptide
Proteins and Peptides
Molecular Formula: C119H182N34O31
Formula Weight: 2584.90
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty