Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-D-Galacturonic acid monohydrate 97%

Supplier: Thermo Scientific Chemicals
Synonyms: D-Galacturonic acid monohydrate(2S,3R,4S,5R)-2,3,4,5-Tetrahydroxy-6-oxohexanoic acid hydrate

J66282-06 J66282-14
10815-488EA 172.22 USD InStock
10815-488 10815-486
α-D-Galacturonic acid monohydrate 97%
alpha-D-Galacturonic acid monohydrate
D-(+)-Galacturonic acid monohydrate can be used in the synthesis of N-(D-galacturonoyl) amino acids and dipeptides.


Formula: C₆H₁₀O₇.H₂O
MW: 212.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00071585
CAS Number: 91510-62-2
EINECS: 211-682-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty