Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Fructose-6-phosphate disodium salt 95%

Supplier: Thermo Scientific Chemicals
Warning

J66311-03 J66311-MC J66311-ME
10815-538EA 136.92 USD InStock
10815-538 10815-540 10815-542
D-Fructose-6-phosphate disodium salt 95%
D-Fructose-6-phosphate disodium salt


Formula: C₆H₁₁Na₂O₉P
Storage Temperature: Freezer
MDL Number: MFCD00135619
CAS Number: 26177-86-6
EINECS: 247-505-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty