Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Mannosamine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: D-Mannosamine hydrochloride

J66316-03 J66316-ME
10815-554EA 145.04 USD InStock
10815-554 10815-552
D-Mannosamine hydrochloride 98%
D-Mannosamine hydrochloride


Formula: C6H14ClNO5
MW: 215.632 g/mol
Melting Pt: 168 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00064557
CAS Number: 5505-63-5
EINECS: 226-847-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty