Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-O-Isopropylidene-D-glyceraldehyde 50% (w/w) in dichloromethane

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxaldehyde(R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde(4R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde

J66337-09 J66337-03 J66337-06
10815-576EA 250.58 USD InStock
10815-576 10815-572 10815-574
2,3-O-Isopropylidene-D-glyceraldehyde 50% (w/w) in dichloromethane
2,3-O-Isopropylidene-D-glyceraldehyde


Formula: C₆H₁₀O₃
MW: 130.143 g/mol
Flash Pt: 62 °C(144 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00269682
CAS Number: 15186-48-8
UN: 1593
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty