Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Galacto-D-mannan

Supplier: Thermo Scientific Chemicals
Synonyms: CarobgalactomannanD-Galacto-D-mannaneGalactomannoglycan

J66610-ME J66610-MC
10815-718EA 123.74 USD InStock
10815-718 10815-720
D-Galacto-D-mannan
D-Galacto-D-mannan


Formula: C₁₈H₃₂O₁₆
MW: 504.437 Da
Storage Temperature: Ambient
MDL Number: MFCD00146683
CAS Number: 11078-30-1
EINECS: 234-299-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty