Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium glucoheptonate dihydrate ≥98.5%

Supplier: Thermo Scientific Chemicals
Synonyms: Glucoheptonic Acid Sodium SaltGlucosecarboxylic Acid Sodium Salt

J66811-22 J66811-36 J66811-A1
10815-848EA 45.5 USD InStock
10815-848 10815-858 10815-850
Sodium glucoheptonate dihydrate ≥98.5%
Sodium glucoheptonate dihydrate


Formula: C₇H₁₄O₈
MW: 248.16 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00013425
CAS Number: 31138-65-5
EINECS: 250-480-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty