Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-D-Thioglucose sodium salt 97%

Supplier: Thermo Scientific Chemicals

J66838-03 J66838-MD
10815-890EA 46.55 USD InStock
10815-890 10815-888
β-D-Thioglucose sodium salt 97%
beta-D-Thioglucose sodium salt


Formula: C₆H₁₁NaO₅S
MW: 218.199 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00061630
CAS Number: 10593-29-0
EINECS: 234-200-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty