Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Di-O-acetyl-6-deoxy-L-glucal 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 3,4-Di-O-acetyl-1,5-anhydro-2,6-dideoxy-L-arabino-hex-1-enitol3,4-Di-O-acetyl-L-rhamnal

J66944-06 J66944-03
10815-916EA 46.51 USD InStock
10815-916 10815-918
3,4-Di-O-acetyl-6-deoxy-L-glucal 98%
3,4-Di-O-acetyl-6-deoxy-L-glucal


Formula: C₁₀H₁₄O₅
MW: 214.22 g/mol
Boiling Pt: 68…69 °C
Density: 1.116
Flash Pt: >110 °C(235 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00074970
CAS Number: 34819-86-8
EINECS: 252-228-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty