Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Gulonic acid-γ-lactone 98%

Supplier: Thermo Scientific Chemicals
Synonyms: D-(-)-Gulono-1,4-lactoneD-Gulono γ-lactoneD-Gulono-1,4-lactone(3R,4S,5S)-5-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one

J66987-06 J66987-14
10816-034EA 206.97 USD InStock
10816-034 10816-032
D-Gulonic acid-γ-lactone 98%
D-Gulonic acid-γ-lactone


Formula: C₆H₁₀O₆
MW: 178.14 g/mol
Melting Pt: 182…188 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005391
CAS Number: 6322-07-2
EINECS: 228-678-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty