Would you like to log in again?
Insterprep® Fraser Broth, Seward
Supplier: Seward
INS-2202 INS-2302 INS-2102
10818-618PK 1841.54 USD InStock
10818-618 10818-616 10025-022
Insterprep® Fraser Broth, Seward
Media for Microbiology Prepared Media for Microbiology
Insterprep® is a new range of instantly soluble, sterile, granulated media specifically designed to speed up and simplify sample preparation for microbiological analysis
These instantly soluble, sterile Fraser Broth media granules are supplied ready to use. Choose the Insterprep® sachet to fit specific protocol needs, or the Stomacher® bag. Simply add sterile distilled water and the sample as per protocol.
The Insterprep® pre-weighed, sterilized media bags are packaged with the correct amount of granular media, ready for instant use. The Insterprep® range of products is designed to help forward thinking laboratories cut waste and costs, manage laboratory resources more efficiently, and produce dependable and accurate results.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty