Would you like to log in again?
Pneumocystis jiroveci Control Slide, Polysciences
Supplier: Polysciences
22251-1
10818-626BX 778.85 USD InStock
10818-626
Pneumocystis jiroveci Control Slide, Polysciences
Slides Microscopy Control Slides
Designed to be used with the Fungi Fluor Pneumocystis Kit
Slides are packaged 10 per box.
Unstained slides feature rat lung imprints and are methanol fixed. Store at room temperature. These control slides are compatible with other staining processes for P. Carinii, including Methenamine Silver Staining and Gram-Weigert techniques.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty