Would you like to log in again?
Virgo® Cardiolipin ELISA Test System, Hemagen
Supplier: Hemagen Diagnostics
Virgo®
66032 66013 66031 66014 66016 66033 66015 66030
10818-784KT 765.2 USD InStock
10818-784 10818-782 10818-794 10818-780 10818-792 10818-790 10818-788 10818-786
Virgo® Cardiolipin ELISA Test System, Hemagen
Assays ELISAs
An enzyme-linked immunosorbent assay (ELISA) designed for the quantitative measurement of circulating IgG, IgA and IgM autoantibodies to cardiolipin.
  • Assay Kit
  • FDA Compliant
  • For quantitative measurement
Certifications: FDA Compliant
Ordering information: Accessories included.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty