Would you like to log in again?
Caps for 12-425 Screw-Thread Vials
Supplier: Thermo Scientific
12-SCS
10820-602PK 108.74 USD InStock
10820-602
Caps for 12-425 Screw-Thread Vials
Closures Chromatography Vial Closures
Screw thread caps feature a 13 to 425 thread finish and are available in an assortment of styles to match your HPLC, GC or IC application needs.
  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Phenolic resin caps perform well at high temperatures and are compatible with exposure to corrosives
  • Caps are bonded septa resist dislodging during injection when using large diameter blunt needles
Convenient pre-assembled caps and septa minimize contamination from handling.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty