Would you like to log in again?
Mag-Bind® Universal Pathogen DNA Kit, Omega Bio-Tek
Supplier: Omega Bio-Tek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Mag-Bind®
M4029-01 M4029-01 M4029-00
10821-236EA 391.86 USD InStock
10821-236 10821-238 CA10821-238
Mag-Bind® Universal Pathogen DNA Kit, Omega Bio-Tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The MagBind Universal Pathogen 96 Kit allows rapid and reliable isolation of high-quality host genomic DNA, gram positive and negative bacterial DNA, fungal spore DNA, viral DNA, and viral RNA from tissue, urine, serum, and fecal samples
  • No organic extractions
  • Pure DNA for downstream applications such as qPCR and next generation sequencing (NGS)
  • Isolate DNA and viral RNA from a wide variety of sample types, including feces
The system allows for automation after sample lysis via Hamilton STAR™, Thermo KingFisher™ Flex, Applied Biosystems® MagMAX™, Qiagen BioSprint, and other liquid handling instruments.
E-Z 96 Disruptor Plus plates are included with the kit for simultaneous homogenization and lysis of the sample. The unique buffer system does not require alcohol to bind nucleic acids allowing for isolation of high-quality DNA/RNA, free of PCR inhibitors. The entire procedure can be completed in less than 60 minutes for 96-samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty