Would you like to log in again?
pH Electrodes, LE427 / LE438, Mettler-Toledo
Supplier: Mettler-Toledo
30259840 30247153 30259840 30247153
10821-384EA 303 USD InStock
10821-384 10821-386 CA10821-384 CA10821-386
pH Electrodes, LE427 / LE438, Mettler-Toledo
Electrodes pH Electrodes
Waterproof pH electrodes for use with portable FiveGo™ meters.
LE438: 3-in-1 plastic pH electrode with integrated temperature sensor for pH field use, samples with fluctuating temperatures; low-maintenance.
LE427: The spear tip design makes pH electrode ideal for solid samples such as meat, fruit, cheese, soil and many more; low-maintenance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty