Would you like to log in again?
GenCollect™ Sample Card, Ahlstrom
Supplier: Ahlstrom
GenCollect™
8.560.0000.B-N 8.560.0002.B-N 8.560.0001.B-N
10821-454PK 364.52 USD InStock
10821-454 10821-456 10821-458
GenCollect™ Sample Card, Ahlstrom
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Biological Sample Collection/Preparation
Ahlstrom's GenCollect™ Sample Collection Card for collection and short-term storage of biological samples at ambient temperature contains untreated paper suitable for storage of whole blood and similar specimens
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty