Would you like to log in again?
Analytical and Precision Balances, ME-TE
Supplier: Mettler-Toledo
ME-T
30216559 30216552 30216566 30216620 30243386 30216563 30216623 30216549 30243381 30216561 30216623 30216566 30216625 30216563 30216557 30216568 30216546 30216549 30216625 30216546 30243381 30243386 30216561 30216568 30216559 30216552 30216557 30216620
76045-426EA 3220 USD InStock
CA76045-596 CA10821-508 76045-426 10821-512 CA10821-520 10821-514 CA10821-500 10821-516 10821-518 CA10821-492 CA10821-494 10821-520 CA10821-496 CA10821-518 10821-508 CA76045-426 10821-500 CA10821-510 CA10821-498 CA10821-512 CA10821-514 76045-596 CA10821-516 10821-492 10821-494 10821-496 10821-498 10821-510
Analytical and Precision Balances, ME-TE
Balances and Scales
METTLER TOLEDO ME-TE balances are reliable balances for every lab that combine essential weighing functions with ease of use to deliver accurate results day after day.
  • Capacities from 52 to 4200 g
  • Readabilities from 0.1 mg
The unit’s large 4.5” color TFT touchscreen with clear icons and intuitive guidance enables users to operate it quickly - even while wearing cloth or rubber gloves.
An Electromagnetic Force Compensation (EMFC) weighing cell delivers consistently reliable results and built-in overload protection helps to ensure a long balance lifetime. The metal base housing and a high quality finish make the ME-TE robust enough for tough environments, while smooth surfaces make it easy to maintain and clean.
Eight embedded applications including checkweighing, totaling and formulation allow operators to start working right away. Sample IDs and passcode protection make sure settings are not changed by unauthorized personnel. Multiple interfaces support barcode reader, printer and PC connections for easy traceability and data handling. USB and RS232 connectivity along with an optional Bluetooth adapter simplify data upload and transfer.
Certifications: UL listed and CSA certified.
Ordering information: The .01g and 0.1g models do not include draft shields. Optional Service packages are offered to assure continual reliability and maintain optimum balance performance. Proper installation and initial calibration are the key to ensuring the balance performs properly from day one. A METTLER TOLEDO technician performs installation qualification and operational qualification (IQ/OQ). Contact your representative for Service package details.
Order Now
ORDER
SPECIFICATIONS
SERVICES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty