Would you like to log in again?
LD-100 System Frocks, 469 Series, WORKLON®
Supplier: Superior Uniform Group
Worklon®
469-3XL 469-XS 469-L 469-XL 469-4XL 469-5XL 469-M 469-2XL 469-S
10827-194EA 146.33 USD InStock
10827-194 10830-610 10827-196 10827-192 10830-608 10827-202 10827-204 10827-198 10827-200
LD-100 System Frocks, 469 Series, WORKLON®
Workwear
Reusable frocks feature a snap closure and set-in sleeves.
  • Adjustable snaps at wrist and collar
  • Color-coded hang loop denotes size
  • Unisex design
The 100% polyester taffeta construction allows for use in a variety of industries requiring particulate control and resistance to chemicals and abrasion. These frocks are engineered to meet up to Class 1000 (ISO 6) cleanroom requirements.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty