Would you like to log in again?
VWR® Pipette Carousel Stand
Supplier: VWR International
VWR®
VWR-CAR
10827-940EA 137.69 USD InStock
10827-940
VWR® Pipette Carousel Stand
Pipette Stands
Rotary pipette stand for up 7 pipettors. Made from PP/PC.
  • Pipettes hang vertically to prevent liquids from running inside the shaft
  • Rotating head for easy access
  • Designed for single and multichannel models
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty