Would you like to log in again?
Whatman™ Pure Quartz Air Sampling Filter, Grade QM-H, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
1853-150-50 1853-150-50 1853-050-50 1853-047-50 1853-047-50 1853-037-50 1853-090-50 1853-090-50 1853-050-50 1853-037-50
10828-404PK 250.08 USD InStock
10828-404 CA10828-402 CA10828-400 10828-402 CA10827-974 CA10828-404 10827-974 10828-400 CA10828-398 10828-398
Whatman™ Pure Quartz Air Sampling Filter, Grade QM-H, Whatman products (Cytiva)
Filters Glass Fiber Filters
Whatman™ QM-H is a pure quartz fibre filter for use in testing acidic gases, stacks, flues and aerosols. It has low heavy metal content so it is well suited for heavy metals analysis.
  • High stability up to 900 °C
  • High filtration efficiency
  • Binder-free
  • Minimal reactivity with biological or chemical components
Pure quartz fiber filters come in 50-piece packs with five diameters: 37 mm, 47 mm, 50 mm, 90 mm, 150 mm.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty