Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ Grade 4V Qualitative Filter Papers, Whatman products (Cytiva)

Supplier: Whatman products (Cytiva)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Whatman™
1204-125 1204-240 1204-240 1204-270 1204-320 1204-150 1204-270 1204-0185 1204-320 1204-125 1204-150 1204-0185
10827-976PK 40.78 USD InStock
CA10827-978 10827-976 10827-986 CA10827-984 CA10827-976 10827-978 CA10827-986 10827-982 CA10827-980 CA10827-982 10827-984 10827-980
Whatman™ Grade 4V Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Whatman Grade 4V filter paper is well suited for qualitative analysis or general filtration of coarse particles. The fluted filter design increases filtration speed, flow rate, and loading capacity.

  • Very fast flow rate
  • Retention of coarse and gelatinous precipitates
  • Nominal particle retention rating of 20 to 25 µm
  • Fast filtration speed (Herzberg 37 seconds)
  • Fluted (pre-pleated) for compatibility with conical filter funnels without the need for further folding

Whatman Grade 4V prepleated (fluted) filter paper from Cytiva is frequently used to filter gelatinous precipitate or during general filtration that does not require the collection of fine particles. The filter sheets are also useful for rapid filtration of biological fluids or organic extracts prior to analysis.

Typical Trace Element Levels: Aluminium 3.6 µg/g Antimony <0.5 µg/g Arsenic <0.5 µg/g Barium <0.5 µg/g Boron <1 µg/g Calcium 27.5 µg/g Chromium 1 µg/g Copper 0.9 µg/g Iron 13.7 µg/g Lead <0.5 µg/g Magnesium 21 µg/g Manganese <0.5 µg/g Mercury <0.5 µg/g Potassium 6.2 µg/g Silicon 8.8 µg/g Sodium 32.3 µg/g Zinc 58.3 µg/g

Water flow rate: 247 ml per minute

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty