Would you like to log in again?
3 COMPARTMENT DISP W/FRONT ACCESS
Supplier: S-CURVE TECHNOLOGIES
Dispenser 3 Compartment With Front Access^ Dispensers, with 3 individual Compartment -Round Front Access Hole/opening (3.5v4inch), 1/4 inch Clear Material: Acrylic, Mounts to wall or free standing, Flame treated edges for a smooth finish, Dimension (WXHXD): 22.5X14X9Inch
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
3 COMPARTMENT DISP W/FRONT ACCESS 10828-386 MCD-350 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty