Would you like to log in again?
5049 Series ESD Jacket/Lab Coat, Transforming Technologies
Supplier: Transforming Technologies
JWC5403SPWH JWC5408SPLB JWC5403WH JWC5405WH JLC5409SPWH JWC5405LB JWC5409SPWH JWC5403LB JWC5407WH JLC5403SPWH JWC5402SPLB JWC5406LB JLC5408SPWH JWC5405SPWH JLC5402SPWH JLC5407SPWH JWC5404LB JWC5403SPLB JWC5406SPLB JLC5406SPLB JLC5409SPLB JWC5406WH JWC5408LB JLC5405SPLB JWC5409WH JWC5407SPWH JLC5408SPLB JWC5404SPLB JLC5404SPLB JLC5402SPLB JWC5402LB JWC5404SPWH JWC5409SPLB JLC5404SPWH JLC5403SPLB JWC5402WH JWC5409LB JLC5405SPWH JWC5407SPLB JWC5405SPLB JLC5406SPWH JWC5406SPWH JWC5407LB JLC5407SPLB JWC5408SPWH JWC5402SPWH JWC5404WH JWC5408WH
10828-900EA 74.17 USD InStock
10828-900 10828-844 10829-190 10828-820 10829-192 10828-948 10829-092 10829-172 10829-150 10828-868 10828-626 10828-682 10828-680 10828-940 10829-108 10828-908 10829-064 10829-184 10829-162 10829-066 10829-164 10829-186 10828-828 10829-144 10829-100 10829-146 10828-812 10828-876 10828-732 10829-060 10829-180 10829-084 10828-836 10828-598 10828-730 10829-116 10829-118 10829-152 10829-174 10829-074 10829-110 10829-076 10828-916 10829-112 10829-178 10829-156 10829-158 10829-058
5049 Series ESD Jacket/Lab Coat, Transforming Technologies
Workwear
The 5049 series ESD jackets are made with more cotton than common polyester ESD jackets which makes the jackets more comfortable, breathable and softer. 5049 Fabric is made with 66% Polyester, 32% Cotton and 2% Carbon.
  • Garment maintains consistent continuity readings for up to 100 wash cycles
  • Breathable cotton poly fabric blend allows for more airflow, and is an excellent choice for very warm climates
  • Reliable panel-to-panel continuity which eliminates the danger of isolated conductors
  • Available in different lengths
  • Available with different cuff options (knitted cuff or hemmed sleeve with three snaps that adjusts for a proper fit)
Perfect for all working environments such as Electronics Manufacturing & Repair, Laboratory, Automotive, Aerospace and General Industrial Applications.
Full length option: A lab coat that goes to knee length - covers more clothing than traditional jackets.
Certifications: Heavyweight Cotton ESD Jacket are all tested and proven to pass ANSI ESD S20.20.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty