Would you like to log in again?
8812 Series ESD Jacket, Transforming Technologies
Supplier: Transforming Technologies
JKV8827BG JKV8827SPBG JKC8805BG JKC8804BG JKV8827LB JKV8826BG JKV8825LB JKC8807LB JKV8822WH JKC8802WH JKV8825SPLB JKV8827WH JKC8807SPBG JKC8802SPWH JKV8824WH JKV8824LB JKV8829SPWH JKC8805SPBG JKC8805LB JKC8809SPBG JKC8808WH JKV8829WH JKV8824SPLB JKC8806LB JKV8828SPBG JKC8808BG JKV8825BG JKC8808SPBG JKV8825SPWH JKC8803LB JKV8826SPBG JKC8809WH JKV8829LB JKV8827SPWH JKV8826WH JKC8803SPLB JKV8825SPBG JKV8829BG JKV8826SPLB JKC8802BG JKV8829SPLB JKC8802LB JKV8826LB JKC8805WH JKV8824BG JKV8827SPLB JKC8807SPWH JKC8803SPBG JKV8828BG JKV8828SPWH JKC8808SPWH JKV8823SPWH JKV8824SPBG JKV8822SPLB JKC8806SPBG JKC8804SPWH JKV8822LB JKC8806BG JKC8809SPWH JKV8829SPBG JKC8803BG JKC8802SPBG JKC8804SPBG JKV8823BG JKC8807WH JKC8804SPLB JKC8807BG JKC8804WH JKV8823LB JKV8824SPWH JKV8828WH JKC8809SPLB JKC8803WH JKC8809BG JKV8828SPLB JKC8809LB JKC8804LB JKC8802SPLB JKC8806WH JKV8826SPWH JKV8828LB JKV8823SPLB JKC8805SPLB JKC8805SPWH JKC8807SPLB JKC8803SPWH JKV8823SPBG JKV8823WH JKC8806SPWH JKC8806SPLB JKV8822SPBG JKV8822BG JKC8808SPLB JKV8822SPWH JKC8808LB JKV8825WH
10828-746EA 82.29 USD InStock
10828-746 10828-988 10828-986 10828-744 10829-070 10828-980 10828-984 10828-742 10828-588 10828-740 10828-982 10828-860 10829-028 10828-590 10829-020 10829-140 10829-022 10829-024 10829-026 10828-658 10828-856 10828-976 10828-734 10828-932 10828-854 10828-930 10828-974 10828-738 10828-858 10828-978 10828-736 10828-892 10828-890 10828-852 10828-972 10828-850 10828-970 10829-016 10829-018 10829-052 10829-010 10829-054 10829-012 10829-014 10828-966 10828-922 10828-888 10828-920 10828-964 10828-886 10829-170 10828-926 10828-848 10828-968 10828-924 10829-050 10828-846 10828-880 10828-962 10828-884 10828-642 10828-960 10828-640 10828-882 10829-006 10829-008 10829-042 10829-000 10829-044 10828-928 10829-046 10829-002 10829-166 10829-004 10829-048 10829-168 10828-636 10828-878 10828-998 10828-634 10828-996 10829-040 10828-638 10828-672 10828-594 10828-990 10828-670 10828-592 10828-994 10828-992 10829-038 10829-030 10828-918 10829-032 10829-034 10829-036
8812 Series ESD Jacket, Transforming Technologies
Workwear
The 8812 Series jackets from Transforming Technologies feature a patented hip-to-cuff feature for hands-free grounding
These garments are fabricated from lightweight 88% polyester and 12% carbon fabric for unmatched operator comfort. Superior workmanship provides reliable panel to panel continuity. The 8812 Series jackets maintains consistent continuity readings for up to 100 wash cycles.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty