Would you like to log in again?
VWR® Benchtop General Purpose Centrifuges
Supplier: VWR
1406-CELLCULT 1401-CELLCULT 1406-37 1406-MICRO 1401-37 1406-PLATE 1401-MICRO 1401-PLATE 1401-PCR 1406-PCR
10830-776EA 12368.46 USD InStock
10830-776 10830-764 10830-774 10830-762 10830-772 10830-770 10830-746 10830-768 10830-744 10830-766
VWR® Benchtop General Purpose Centrifuges
Centrifuges Benchtop Centrifuges
This compact benchtop unit offers laboratories a true workflow benefit due to its vast capabilities as mid-sized, general purpose, tabletop centrifuge.
  • Offers a unique fast-cool function
  • General-purpose use
  • Mid-sized tabletop centrifuge is compatible with most laboratory workflows
Available with temperature control, the refrigerated unit offers a unique fast-cool function that will lower the unit to +4 °C in 10-15 minutes saving you valuable time spinning delicate samples.
Whether you are working with blood, urine, cell/tissue culture, plates or cytology the compact benchtop centrifuge will provide consistent, accurate results spin after spin. For additional peace of mind, VWR® centrifuges come with a Five (5)-year warranty. For quick and easy ordering be sure to view VWR® centrifuge packages. The centrifuge bundles ship complete with all the necessary components to perform your applications effortlessly.
Ordering information: Units come with a five-year warranty that extends the standard two-year warranty for an additional three years. The three-year extension is a full powertrain warranty on the centrifuge only.
Order Now
ORDER
ACCESSORIES
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty