Would you like to log in again?
OFX-100 Lab Coats, Tech Wear
Supplier: Tech Wear
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LOC-23-XS LOC-23-M LOC-23-2XL LOC-23-4XL LOC-23-S LOC-23-5XL LOC-23-L LOC-23-XL LOC-23-3XL
10831-928EA 411.23 USD InStock
10831-928 10831-918 10831-924 10831-934 10831-926 10831-920 10831-930 10831-922 10831-932
OFX-100 Lab Coats, Tech Wear
Workwear
Knee-length lab coats are made of OFX-100 static control fabric and maintain sleeve-to-sleeve continuity for a minimum of 100 washes or two years, whichever comes first
They feature a lapel collar and three pockets. Lab coats are conductive and meet ANSI/ESD STM2.1-1997 garment requirements.
Ordering information: Lab coats are also available grounded and in white and teal; contact your Avantor sales representative for information.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty