Would you like to log in again?
Lightweight OFX-100 Jackets, Tech Wear
Supplier: Tech Wear
LOJ-23-3XL LOJ-23-XS LOJ-23-4XL LOJ-23-S LOJ-23-M LOJ-23-2XL LOJ-23-5XL LOJ-23-L LOJ-23-XL
10831-940EA 308.6 USD InStock
10831-940 10831-950 10831-938 10831-946 10831-948 10831-936 10831-942 10831-952 10831-944
Lightweight OFX-100 Jackets, Tech Wear
Workwear
Jackets are 87% polyester and 13% conductive BASF Resistat® fiber, which has a static decay rate of 5000 to 500V in less than 0.1 seconds.
Hip-length jackets feature a lapel collar and three pockets. They meet ANSI/ESD STM2.1-1997 requirements.
Ordering information: Jackets are also available grounded and in white and burgundy; contact your VWR sales representative for information.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty