Would you like to log in again?
Cleanroom Boot Covers made with DuPont™ Tyvek® Material, HPK Industries
Supplier: HPK INDUSTRIES LLC
52447K-XL 52447K-M/L
10832-142CS 2054.86 USD InStock
10832-142 10832-146
Cleanroom Boot Covers made with DuPont™ Tyvek® Material, HPK Industries
Disposable Garments
Boot covers have adjustable ties and elastic tops.
  • PVC bottoms provide durability and skid-resistance
  • Bound seam construction
  • Sterile
For custom sizes and styles, contact your Avantor sales representative.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty