Would you like to log in again?
Alliance Sterile Coveralls, HPK Industries
Supplier: HPK Industries
151-S 151-XL 151-XXL 151-L 151-M
10832-176CS 1440.78 USD InStock
10832-176 10832-162 10832-178 10832-160 10832-180
Alliance Sterile Coveralls, HPK Industries
Disposable Garments
An extremely smooth outer fabric surface lowers particle cling and build-up in cleanroom applications
Fabric is wrinkle-free and highly durable. Built-in “cooling system” dissipates heat and moisture for high worker comfort. Elastic wrists and ankles. Bound seam construction. Sterile. Hood and boot covers (shown) not included.
Individually packaged in a sealed bag.
Coverall material acts as a liquidproof barrier. Unique antistatic system assures static dissipation in sensitive cleanroom work areas.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty