Would you like to log in again?
Alliance Visitor Kits, HPK Industries
Supplier: HPK Industries
475-L 475-XXL 475-S 475-XL 475-M
10832-198CS 2692.62 USD InStock
10832-198 10832-196 10832-190 10832-194 10832-192
Alliance Visitor Kits, HPK Industries
Disposable Garments
For contamination control in the cleanroom
Zipper-front coveralls feature elastic wrists and ankles. Hood has adjustable snap crown for proper fit. Calf-high boots have nonskid bottom.
Visitor kit consists of coveralls, hood and boots constructed of liquidproof, breathable fabric. Microporous material dissipates heat and moisture, cooling the body. Antistat system assures static dissipation.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty