Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BD BBL™ Streptocard™ Enzyme Latex Test Kits, BD Diagnostics

Supplier: BD Diagnostics

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

BD BBL™
240957 240928 240935 240918 240933 240958 240924 240956 240938
10834-118EA 135.66 USD InStock
10834-118 10834-128 10834-116 10834-048 10834-124 10834-122 10834-120 10834-130 10834-040
BD BBL™ Streptocard™ Enzyme Latex Test Kits, BD Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
The BD BBL™ Streptocard™ enzyme latex test kits, provides a rapid platform for the serological identification of beta-haemolytic streptococci belonging to Lancefield groups A, B, C, D, F and G.

  • Materials are supplied ready for use
  • All kit components should be stored at 2 to 8 °C
  • Do not freeze
  • For in vitro diagnostic use

The grouping method involves enzymatic extraction of group specific carbohydrate antigens. The strep enzyme extraction reagent provided in the kit contains lytic enzymes able to extract the streptococcal group specific antigens with incubation at 37 °C. The extracts can be easily identified using blue polystyrene latex particles sensitized with purified group specific rabbit immunoglobulins. These blue latex particles agglutinate very strongly in the presence of homologous antigen and will not agglutinate when homologous antigen is absent.

BD BBL™ Streptocard™ enzyme latex test kits: Each dropper bottle contains 2.5 ml of blue latex particles coated with purified rabbit antibodies to Lancefield groups A, B, C, D, F or G streptococci. The blue latex particles are suspended in a pH 7.4 buffer containing 0.098% sodium azide as a preservative.

BD BBL™ Streptocard™ enzyme positive control: One dropper bottle containing 2.5 ml of ready to use polyvalent antigens extracted from inactivated streptococci of Lancefield groups A, B, C, D, F and G. The antigens are suspended in a pH 7.4 buffer containing 0.098% sodium azide as a preservative.

BD BBL™ Streptocard™ extraction enzyme: One bottle containing 22 ml of ready to use enzyme extraction reagent with preservative.

Delivery information: Test cards, mixing sticks, instructions for use will be supplied.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty