Would you like to log in again?
TB Base Digestant, Hardy Diagnostics
Supplier: Hardy Diagnostics
X45
10834-146PK 182.47 USD InStock
10834-146
TB Base Digestant, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
TB Base Digestant, for the digestion and decontamination of sputum specimens for TB culture
  • For the processing of specimens for AFB culture.
  • Add NALC upon use
For the processing of specimens for AFB culture
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.
Add NALC upon use.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty