Would you like to log in again?
Human Recombinant Active Cathepsin S (from E. coli)
Supplier: BioVision
AB285884-10UG AB285884-500UG AB285884-50UG
10834-684EA 13341.31 USD InStock
10834-684 10834-682 10834-680
Human Recombinant Active Cathepsin S (from E. coli)
Proteins and Peptides
≥9% pure active human recombinant Cathepsin S
Cathepsin S (CTSS) is a lysosomal cysteine protease of the papain family and may participate in the degradation of antigenic proteins to peptides for presentation on MHC class II molecules. CTSS is synthesized as inactive precursor of 331 amino acids consisting of a 15-aa signal peptide, a propeptide of 99 aa, and a mature polypeptide of 217 aa. It is activated in the lysosomes by a proteolytic cleavage of the propeptide. The deduced amino acid sequence contains only one potential N-glycosylation site located in the propeptide. Compared with the abundant cathepsins B, L and H, cathepsin S shows a restricted tissue distribution, with highest levels in spleen, heart, and lung. In addition, evidences indicate that cathepsin S generates amyloid beta-peptide from amyloidogenic fragments of amyloid precursor protein (APP) in the endosomal/lysosomal compartment, and is implicated in the pathogenesis of Alzheimer’s disease and Down Syndrome.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty