Would you like to log in again?
Human Recombinant Enterokinase Light Chain (from E. coli)
Supplier: BioVision
AB285887-500UG AB285887-10UG AB285887-50UG
10834-696EA 1478.53 USD InStock
10834-696 10834-694 10834-692
Human Recombinant Enterokinase Light Chain (from E. coli)
Enzymes
≥9% pure active human recombinant Enterokinase (Light Chain)
Enterokinase (EC 3.4.21.9) is a serine protease involved in the activation of trypsinogen to trypsin, which in turn results in the activation of various digestive enzymes. It recognizes a highly specific amino acid sequence ‘DDDDK’ and cleaves after the lysine residue. High specific activity of enterokinase has been utilized in cleaving a variety of native or fusion protein tags containing the above recognition motif.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty