Would you like to log in again?
Human Recombinant Active Cathepsin B (from E. coli)
Supplier: BioVision
AB285903-50UG AB285903-5UG AB285903-1MG
10834-768EA 23105.06 USD InStock
10834-768 10834-766 10834-764
Human Recombinant Active Cathepsin B (from E. coli)
Proteins and Peptides
>9% Pure active human recombinant Cathepsin B.
  • >7500 mU/mg (1U = 1 µmole/min digestion of Ac-Arg-Arg-AFC substrate (K140-100)).
Cathepsin B is a lysosomal cysteine protease. It is implicated in playing a role in protein degradation, arthritis, apoptosis, and cancer.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty