Would you like to log in again?
Human Recombinant Glycine Oxidase H244K Bs (from E. coli)
Supplier: BioVision
AB285972-1MG AB285972-250UG
10835-720EA 4067.92 USD InStock
10835-720 10835-718
Human Recombinant Glycine Oxidase H244K Bs (from E. coli)
Enzymes
≥9% Pure Active Bacillus subtilis recombinant Glycine Oxidase H244K
1200 mU/mg
Glycine oxidase (GO) from Bacillus subtilis (EC 1.4.3.19) is a homotetrameric flavin-dependent oxidoreductase. Each GO monomer is non-covalently bound to flavin adenine dinucleotide. GO catalyzes oxidative deamination of various primary and secondary amines (e.g. glycine, sarcosine, N-ethylglycine) and some D-amino acids (e.g. D -alanine, D -proline, D -valine) to the corresponding α-keto acids and hydrogen peroxide. Primarily, glycine oxidase catalyzes the oxidation of glycine in the biosynthesis of thiamine. The variant H244K shows a higher substrate specificity ratio for glycine versus sarcosine and a 5-fold improved specific activity in comparison to the wild-type.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty