Would you like to log in again?
Human Recombinant Active SIRT7 (from E. coli)
Supplier: BioVision
AB285974-10UG AB285974-50UG
10835-722EA 398.24 USD InStock
10835-722 10835-724
Human Recombinant Active SIRT7 (from E. coli)
Proteins and Peptides
≥85% Pure Active Recombinant Human SIRT7
BioVision’s Active His-tagged SIRT7 has been tested for its activity using Sirtuin Activity Assay Kit (K324-100). It has a specific activity of >1 µU/mg.
The sirtuins represent a distinct class of trichostatin A-insensitive lysyl-deacetylases (class III HDACs) and have been shown to catalyze a reaction that couples lysine deacetylation to the formation of nicotinamide and O-acetyl-ADP-ribose from NAD+ and the abstracted acetyl group. There are seven human sirtuins, which have been designated SIRT1-SIRT7. Recently, SIRT7 has been shown to activate transcription by RNA polymerase I and deacetylate p53. SIRT7 prevents progressive functional deterioration of the heart, and is suggested to play an important role in regulation of stress responses and cell death in the heart.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty