Would you like to log in again?
CERTIREF EUP
Supplier: Mettler Toledo
CertiRef* EUP Simplify Performance Verification^ CertiRef* EUP Simplify Performance Verification, Include 7certified reference substance solution & respective blank solvent to perform automatic calibration according to Ph.Eur regulation, for resolution, photometric, wavelength accuracy & stray light
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CERTIREF EUP 10835-912 30254732 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty