Would you like to log in again?
FLOW CELL QUARTZ GLASS 10MM
Supplier: Mettler Toledo
Flow cell, 10 mm^ Flow cell, 10 mm pathlength, 440 uL volume, Suprasil Quartz glass, 170-2700 nm, M6 connectors, PTFE lid
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FLOW CELL QUARTZ GLASS 10MM 10835-920 30258741 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty