Would you like to log in again?
VWR® Pop Up Razor Blade Dispenser and Razor Refill Cartridges
Supplier: Avantor
VWR®
84-0380-0000
10835-969CS 428.9 USD InStock
10835-969
VWR® Pop Up Razor Blade Dispenser and Razor Refill Cartridges
Blade Removers and Dispensers
The VWR® Pop Up Razor Blade Dispenser offers easy dispensing with safe blade storage.
  • Reduce injuries
  • Heavy-duty design with nonskid, heavy bottom
  • Flip-open front enables easy cartridge loading and removal
  • In-use blade holder included
Razor Refill Cartridges are supplied cleanroom ready in clamshell packaging.
The VWR® Pop Up Razor Refill Cartridges are packaged 10 cartridges per case. Each cartridge is labeled with a blade type detail ("CS" for carbon steel blades, "SB" for steel back blades, "W" for washed blades, or "3" for 3 facet blades). The clamshell packaging is cleanroom ready.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty