Would you like to log in again?
VWR® Razor Blade Cartridges for Pop-Up Dispenser
Supplier: Avantor
VWR®
84-0375-0000 84-0374-0000 84-0377-BULK 84-0371-0000 84-0378-0000 84-0376-0000 84-0379-0000 84-0372-0000 84-0370-0000 84-0379-BULK
10835-963CS 667.77 USD InStock
10835-963 10835-977 76209-670 10835-965 10835-971 10835-981 10835-973 76209-668 10835-979 10835-967
VWR® Razor Blade Cartridges for Pop-Up Dispenser
Razors and Blades
Razor Refill Cartridges for use with the VWR® Pop-Up Dispenser.
Razor Refill Cartridges are supplied cleanroom ready in clamshell packaging.
The VWR® Pop Up Razor Refill Cartridges are packaged 10 cartridges per case. Each cartridge is labeled with a blade type detail ('CS' for carbon steel blades, 'SB' for steel back blades, 'W' for washed blades, or '3' for 3 facet blades).
Ordering information: Dispenser and cartridges have to be ordered separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty