Would you like to log in again?
Microchem® by AlphaTec™ 68-1500 Industrial Coveralls with Serged Seams, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WH15-S-92-100-09 WH15-S-92-101-05 WH15-S-92-100-05 WH15-S-92-100-04 WH15-S-92-101-09 WH15-S-92-100-02 WH15-S-92-100-08 WH15-S-92-106-03 WH15-S-92-106-09 WH15-S-92-100-03 WH15-S-92-106-08 WH15-S-92-101-04 WH15-S-92-106-05 WH15-S-92-101-02 WH15-S-92-101-03 WH15-S-92-106-06 WH15-S-92-106-02 WH15-S-92-100-06 WH15-S-92-106-04 WH15-S-92-101-07 WH15-S-92-106-07 WH15-S-92-101-06 WH15-S-92-101-08 WH15-S-92-100-07
10836-114CS 153.48 USD InStock
10836-114 10836-136 10836-112 10836-134 10836-122 10836-100 10836-118 10836-106 10836-128 10836-138 10836-116 10836-104 10836-126 10836-108 10836-090 10836-094 10836-092 10836-132 10836-110 10836-098 10836-120 10836-130 10836-096 10836-140
Microchem® by AlphaTec™ 68-1500 Industrial Coveralls with Serged Seams, Ansell
Disposable Garments
These MICROCHEM® by AlphaTec™ 1500 coveralls have been designed for workers involved in the stripping, clean-up, or handling of asbestos, as well as general maintenance, construction, and contract cleaning.
  • Air and water vapor permeable to help reduce the risk of heat stress
  • Silicone free which is critical in spray painting applications
  • Filter size of particles >3 microns tested with JSTIIF particle penetration test
  • Optimized body fit improves wearer comfort and safety
Serged seams combine strength with particle barrier capabilities. Garments feature elasticated hood, wrists, and ankles, and a 2-way front zipper with storm flap.
Certifications: Tested according to ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty