Would you like to log in again?
Microchem® by AlphaTec™ 68-1800 Industrial Coveralls with Low Hazard Liquid Protection and Bound Seams, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WH18-B-92-103-04 WH18-B-92-103-07 WH18-B-92-111-02 WH18-B-92-107-07 WH18B9210309 WH18-B-92-107-05 WH18-B-92-111-08 WH18B9210308 WH18-B-92-111-03 WH18-B-92-111-09 WH18-B-92-111-07 WH18-B-92-111-08 WH18-B-92-111-09 WH18-B-92-103-05 WH18-B-92-111-03 WH18-B-92-111-07 WH18-B-92-111-04 WH18-B-92-107-04 WH18-B-92-111-05 WH18-B-92-111-06 WH18-B-92-111-04 WH18-B-92-111-05 WH18-B-92-103-03 WH18-B-92-111-02 WH18-B-92-103-06 WH18-B-92-111-06 WH18-B-92-103-02
10836-146CS 266.46 USD InStock
10836-146 10836-144 10836-348 10836-148 10836-346 10836-150 76498-888 10836-154 10836-472 10836-152 10836-356 10836-476 10836-156 10836-354 10836-474 10836-338 76498-902 76498-900 10836-478 76498-890 76498-898 10836-480 76498-896 76498-894 10836-142 10836-482 76498-892
Microchem® by AlphaTec™ 68-1800 Industrial Coveralls with Low Hazard Liquid Protection and Bound Seams, Ansell
Disposable Garments
MICROCHEM® by AlphaTec™ 1800 coveralls provide the wearer with entry level, low hazard liquid and particulate protection.
  • Low hazard liquid spray, splash, and particle protection
  • Moisture vapor permeable for breathability to help reduce the risk of heat stress
  • Low linting reduces the risk of fiber contamination in some critical areas
  • Antistatic tested according to EN1149-5 and AATCC 76
These disposable latex coveralls with bound seams have a 2-way front zipper with resealable storm flap and elasticated wrists, waist, and ankles. Ideal for a wide range of general industrial applications like general maintenance, paint spraying, powder handling, and food industries.
Certifications: Tested according to EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty