Would you like to log in again?
Microchem® by AlphaTec™ 68-3000 Chemical Protection Coveralls, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
YE30W9212204 YE30W9212209 YE30W9212205 YE30-W-92-111-07 YE30-W-92-111-06 YE30W9212202 YE30-W-92-111-05 YE30-W-92-122-06 YE30W9212203 YE30-W-92-111-03 YE30W9212207 YE30-W-92-111-02 YE30-W-92-111-09 YE30-W-92-111-04 YE30-W-92-111-08
10836-224CS 314.54 USD InStock
10836-224 10836-432 10836-222 10836-232 10836-430 10836-228 10836-436 10836-512 10836-226 10836-434 10836-428 10836-438 10836-218 10836-220 10836-230
Microchem® by AlphaTec™ 68-3000 Chemical Protection Coveralls, Ansell
Disposable Garments
MICROCHEM® by AlphaTec™ 3000 features one of the lightest and most comfortable chemical protective materials on the market today
  • Multi-layer barrier fabric effective against numerous chemicals
  • Bright yellow for improved worker safety
  • Typical coverall features include dual zip systems and double cuffs
  • Antistatic tested according to EN1149-5 and AATCC76
Typical areas of application include chemicals, oil and petrochemicals, pharmaceutical, food industry (caustic clean downs), sewage purification installations, industrial and tank cleaning, and mining.
This durable multilayer fabric provides an extremely effective barrier against both inorganic chemicals and biological hazards.
Certifications: Tested according to EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B, EN ISO 13982-1 Type 5-B, ANSI/ISEA 101.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty